Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST3 Mouse anti-Human, Clone: OTI7G8, liquid, TrueMAB™
SDP

Catalog No. 501677317
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7317 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7317 Supplier OriGene Supplier No. TA600254
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

As an inhibitor of cysteine proteinases, this protein is thought to serve an important physiological role as a local regulator of this enzyme activity.

Specifications

Antigen CST3
Classification Monoclonal
Clone OTI7G8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <5% trehalose and 0.05% sodium azide
Gene CST3
Gene Accession No. P01034
Gene Alias ARMD11, HEL-S-2
Gene Symbols CST3
Host Species Mouse
Immunogen Full length human recombit protein of human CST3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1471
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.