Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CST4 Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504095S


Description
CST4 Monoclonal antibody specifically detects CST4 in Human samples. It is validated for Western BlotSpecifications
CST4 | |
Monoclonal | |
0.91 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WWV3 | |
Mouse | |
Affinity chromatography | |
RUO | |
84816 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2C2 | |
Unconjugated | |
Rtn4ip1 | |
CST4; cystatin 4; cystatin S; cystatin-4; Cystatin-S; cystatin-SA-III; Salivary acidic protein 1 | |
Human recombinant protein fragment corresponding to amino acids 41-279 of human RTN4IP1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction