Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CST8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988912
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8912 Supplier Bioss Supplier No. BS9216R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The cystatin superfamily is a well-established family of cysteine protease inhibitors. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, while some also inhibit legumain family enzymes. The CRES (cystatin-related epididymal spermatogenic) protein defines a new subgroup in the family 2 cystatins of the cystatin superfamily. CRES proteins lack two of the three consensus sites necessary for the cystatin inhibition of C1 cysteine proteases. They are also preferentially expressed in postmeiotic germ cells, the proximal caput epididymidis, and anterior pituitary gonadotrophs. Therefore, CRES proteins may perform unique and tissue-specific functions in the reproductive and neuroendocrine systems. As a member of the CRES subfamily, Cystatin 8 is a 142 amino acid protein that is expressed in the proximal caput region of the epididymis, where it performs a specialized role during sperm development and maturation.

Specifications

Antigen CST8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CST8
Gene Accession No. O60676
Gene Alias Cres; CST 8; Cst8; Cst-rs1; CTES5; cystatin 8; cystatin 8 (cystatin-related epididymal specific); cystatin 8 (cystatin-related epididymal spermatogenic); Cystatin-8; Cystatin-related epididymal spermatogenic protein; Cystatin-related epididymal-specific protein
Gene Symbols CST8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CST8/Cystatin 8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10047
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.