Learn More
Invitrogen™ CSTF1 Monoclonal Antibody (1C6)

Description
Peptide Sequence: LWEISTGRTL VRYTGAGLSG RQVHRTQAVF NHTEDYVLLP DERTISLCCW DSRTAERRNL LSLGHNNIVR CIVHSPTNPG FMTCSDDFRA RFWYRRSTTD.
Specifications
Specifications
| Antigen | CSTF1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1C6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CSTF1 |
| Gene Accession No. | Q05048 |
| Gene Alias | 1700057K18Rik; AI788832; CF-1 50 kDa subunit; cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; Cstf1; CstF50; CstF-50; CstFp50 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.