Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583864 Supplier Invitrogen™ Supplier No. PA583864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGSADASIKI LDTERMLAKS AMPIEVMMNE TAQQNMENHP VIRTLYDHVD EVTCLAFHPT EQILASGSRD YTLKLFDYSK PSAKRAFKYI QEAEMLRSIS Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

This gene encodes one of three subunits which combine to form cleavage stimulation factor (CSTF). CSTF is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. Similar to mammalian G protein beta subunits, this protein contains transducin-like repeats. Several transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSTF1
Gene Accession No. Q05048
Gene Alias 1700057K18Rik; AI788832; CF-1 50 kDa subunit; cleavage stimulation factor 50 kDa subunit; Cleavage stimulation factor subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kD; cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa; CSTF 50 kDa subunit; Cstf1; CstF50; CstF-50; CstFp50
Gene Symbols CSTF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CSTF1. Recombinant protein control fragment (Product #RP-99310).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.