Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSTF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596940 Supplier Invitrogen™ Supplier No. PA596940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGLTVRDPA VDRSLRSVFV GNIPYEATEE QLKDIFSEVG PVVSFRLVYD RETGKPKGYG FCEYQDQETA LSAMRNLNGR EFSGRALRVD NAASEKNKEE LKSLGTGAPV IESPYGETIS PEDAPESISK AVASLPPEQM FELMKQMKLC VQNSPQEARN MLLQNPQLAY ALLQAQVVMR; Positive Samples: HeLa, Jurkat, HepG2, SW620, Mouse testis, Mouse thymus; Cellular Location: Nucleus.

CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3'end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSTF2
Gene Accession No. P33240, Q8BIQ5
Gene Alias alphaCstF-64 variant 4; betaCstF-64 variant 1; betaCstF-64 variant 2; betaCstF-64 variant 3; C630034J23Rik; CF-1 64 kDa subunit; cleavage stimulation factor 64 kDa subunit; Cleavage stimulation factor subunit 2; cleavage stimulation factor, 3' pre-RNA subunit 2; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa; cleavage stimulation factor, 3' pre-RNA, subunit 2; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa; CstF 64; CSTF 64 kDa subunit; Cstf2; Cstf64; CstF-64; mir-3112
Gene Symbols CSTF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human CSTF2 (NP_0013161).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108062, 1478, 683927
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.