Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSTF2T Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101528 Supplier Invitrogen™ Supplier No. PA5101528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CSTF2T.

CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF2T
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSTF2T
Gene Accession No. Q8C7E9, Q9H0L4
Gene Alias 64kDa; C77975; CF-1 64 kDa subunit tau; CF-1 64 kDa subunit tau variant; CF-1 64 kDa subunit, tau; cleavage stimulation factor 64 kDa subunit tau; Cleavage stimulation factor 64 kDa subunit tau variant; cleavage stimulation factor 64 kDa subunit, tau; cleavage stimulation factor subunit 2 tau; cleavage stimulation factor subunit 2 tau variant; cleavage stimulation factor subunit 2, tau variant; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa, tau; cleavage stimulation factor, 3' pre-RNA subunit 2, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant; cleavage stimulation factor, 3' pre-RNA, subunit 2, tau; CSTF 64 kDa subunit tau; CSTF 64 kDa subunit tau variant; CSTF 64 kDa subunit, tau; CSTF2T; CstF-64T; Kiaa0689; TauCstF-64; tCstF-64; Unknown (protein for MGC:129118)
Gene Symbols CSTF2T
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSTF2T(Accession Q9H0L4), corresponding to amino acid residues A93-L143.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23283, 309338, 83410
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.