Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CSTF2T Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541856
Description
Peptide sequence: MRGPVPSSRG PMTGGIQGPG PINIGAGGPP QGPRQVPGIS GVGNPGAGMQ Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 91%; Rat: 85%.
CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.
Specifications
CSTF2T | |
Polyclonal | |
Unconjugated | |
CSTF2T | |
64kDa; C77975; CF-1 64 kDa subunit tau; CF-1 64 kDa subunit tau variant; CF-1 64 kDa subunit, tau; cleavage stimulation factor 64 kDa subunit tau; Cleavage stimulation factor 64 kDa subunit tau variant; cleavage stimulation factor 64 kDa subunit, tau; cleavage stimulation factor subunit 2 tau; cleavage stimulation factor subunit 2 tau variant; cleavage stimulation factor subunit 2, tau variant; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa, tau; cleavage stimulation factor, 3' pre-RNA subunit 2, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant; cleavage stimulation factor, 3' pre-RNA, subunit 2, tau; CSTF 64 kDa subunit tau; CSTF 64 kDa subunit tau variant; CSTF 64 kDa subunit, tau; CSTF2T; CstF-64T; Kiaa0689; TauCstF-64; tCstF-64; Unknown (protein for MGC:129118) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23283 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0L4 | |
CSTF2T | |
Synthetic peptide directed towards the C-terminal of human CSTF2T (aa 460-509). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction