Learn More
Invitrogen™ CSTF2T Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592247
Description
Immunogen sequence: TRAMETEVLE TRVMERRGME TCAMETRGME ARGMDARGLE MRGPVPSSRG PMTGGIQGPG PINIGAGGPP QGPRQVPGIS GVGNPGAGMQ GTGIQGTGMQ GAGIQGGGMQ GAGIQGVSIQ GGGIQGGGIQ GASKQGGSQP S; Positive Samples: Jurkat, U-87MG, K-562, SW620, BT-474, Mouse liver; Cellular Location: Nucleus.
CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.
Specifications
CSTF2T | |
Polyclonal | |
Unconjugated | |
CSTF2T | |
64kDa; C77975; CF-1 64 kDa subunit tau; CF-1 64 kDa subunit tau variant; CF-1 64 kDa subunit, tau; cleavage stimulation factor 64 kDa subunit tau; Cleavage stimulation factor 64 kDa subunit tau variant; cleavage stimulation factor 64 kDa subunit, tau; cleavage stimulation factor subunit 2 tau; cleavage stimulation factor subunit 2 tau variant; cleavage stimulation factor subunit 2, tau variant; cleavage stimulation factor, 3' pre-RNA subunit 2, 64 kDa, tau; cleavage stimulation factor, 3' pre-RNA subunit 2, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau; cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant; cleavage stimulation factor, 3' pre-RNA, subunit 2, tau; CSTF 64 kDa subunit tau; CSTF 64 kDa subunit tau variant; CSTF 64 kDa subunit, tau; CSTF2T; CstF-64T; Kiaa0689; TauCstF-64; tCstF-64; Unknown (protein for MGC:129118) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23283, 83410 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C7E9, Q9H0L4 | |
CSTF2T | |
Recombinant fusion protein containing a sequence corresponding to amino acids 420-560 of human CSTF2T (NP_0560501). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.