Learn More
CSTF2T/TauCSTF64 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301486AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
CSTF2T may play a significant role in AAUAAA-independent mRNA polyadenylation in germ cells. It is directly involved in the binding to pre-mRNAs.Specifications
| CSTF2T/TauCSTF64 | |
| Polyclonal | |
| Unconjugated | |
| CSTF2T | |
| Kiaa0689 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23283 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9H0L4 | |
| CSTF2T | |
| Between 225 and 275 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.