Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSTF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588201 Supplier Invitrogen™ Supplier No. PA588201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGDGATEQA AEYVPEKVKK AEKKLEENPY DLDAWSILIR EAQNQPIDKA RKTYERLVAQ FPSSGRFWKL YIEAEVTILF YFFLYQYCSI HCSDRKQVRN IAN; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.

The protein encoded by this gene is one of three (including CSTF1 and CSTF2) cleavage stimulation factors that combine to form the cleavage stimulation factor complex (CSTF). This complex is involved in the polyadenylation and 3' end cleavage of pre-mRNAs. The encoded protein functions as a homodimer and interacts directly with both CSTF1 and CSTF2 in the CSTF complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CSTF3
Gene Accession No. Q12996, Q99LI7
Gene Alias 4732468G05Rik; C79532; CF-1 77 kDa subunit; cleavage stimulation factor 77 kDa subunit; Cleavage stimulation factor subunit 3; cleavage stimulation factor subunit 3, isoform 1; cleavage stimulation factor, 3' pre-RNA, subunit 3; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa; CSTF 77 kDa subunit; CSTF3; CSTF-77; MGC117398; MGC43001; MGC75122
Gene Symbols CSTF3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human CSTF3 (NP_0010286771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1479, 228410, 362178
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.