Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CT054 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5942 Supplier Bioss Supplier No. BS4164R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a riboflavin transporter protein that is strongly expressed in the intestine and likely plays a role in intestinal absorption of riboflavin. The protein is predicted to have eleven transmembrane domains and a cell surface localization signal in the C-terminus. Mutations at this locus have been associated with Brown-Vialetto-Van Laere syndrome and Fazio-Londe disease. [provided by RefSeq, Mar 2012].

Specifications

Antigen CT054
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC52A3
Gene Accession No. Q9NQ40
Gene Alias bA371L19.1; BVVLS; BVVLS1; C20orf54; hRFT2; RFT2; RFVT3; Riboflavin transporter 2; SLC52A3; solute carrier family 52 (riboflavin transporter), member 3; solute carrier family 52 member 3; Solute carrier family 52, riboflavin transporter, member 3
Gene Symbols SLC52A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CT054/C20orf54.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113278
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.