Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CT110 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$262.00 - $322.50
Specifications
Antigen | CT110 |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CT110 Polyclonal antibody specifically detects CT110 in Human samples. It is validated for Western BlotSpecifications
CT110 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Immunology | |
PBS (pH 7.0) | |
55105 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cancer/testis antigen 110, CT110, FLJ10252, FLJ21048, G patch domain containing 2, G patch domain-containing protein 2, GPATC2, MGC74998, RP11-361K17.1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CT110. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title