Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CT112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560432 Supplier Invitrogen™ Supplier No. PA560432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRLEIYQSSQ DEPIALDKQH SRDSAAITHS TYSLPASSYS QDPVYANGGL NYSYRGYGAL SSNLQPPASL QTGNHSNGP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

C20orf112, also termed as chromosome 20 open reading frame 112, is a 436 amino acid protein, which consists of a relatively hydrophobic N terminal region and two putative small coiled-coil domains. C20orf112 may likely involve in endocytosis pathway and normal brain development. It also is required to mediate the cell energy response. The function of C20orf112 is still non-known and mechanism is required to further study.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CT112
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOL4L
Gene Accession No. Q96MY1
Gene Alias 4931410H15; 8430427H17Rik; AI429157; C20orf112; C20orf113; dJ1184F4.2; dJ1184F4.4; Gm1636; NOL4L; nucleolar protein 4 like; nucleolar protein 4-like; RGD1563510
Gene Symbols NOL4L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CT112. Recombinant protein control fragment (Product #RP-99466).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.