Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CT13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987137
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7137 Supplier Bioss Supplier No. BS6226R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is an ATP-dependent RNA helicase in the DEAD-box family and displays tumor-specific expression.

Specifications

Antigen CT13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DDX43
Gene Accession No. Q9NXZ2
Gene Alias Cancer/testis antigen 13; CT13; DDX43; DEAD (Asp-Glu-Ala-Asp) box polypeptide 43; DEAD box protein 43; DEAD box protein HAGE; DEAD-box helicase 43; DEAD-box protein 43; ENSMUSG00000070291; HAGE; Helical antigen; OTTMUSG00000019690; Probable ATP-dependent RNA helicase DDX43
Gene Symbols DDX43
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DDX43.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55510
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.