Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CT45A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560816 Supplier Invitrogen™ Supplier No. PA560816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGPTAVRKRF FESIIKEAAR CMRRDFVKHL KKKLKRMI Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 45%.

CT45A3 (Cancer/Testis Antigen Family 45 Member A3) is a Protein Coding gene. An important paralog of this gene is CT45A1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CT45A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CT45A5
Gene Accession No. P0DMU7, P0DMU8, P0DMU9, Q5HYN5, Q8NHU0
Gene Alias cancer/testis antigen 45-1; Cancer/testis antigen 45-3; cancer/testis antigen 45-4; cancer/testis antigen 45-4/6; cancer/testis antigen 45-5; Cancer/testis antigen 45-6; cancer/testis antigen 45A1; Cancer/testis antigen 45A10; cancer/testis antigen 45A3; Cancer/testis antigen 45A4; cancer/testis antigen 45A4/45A6; cancer/testis antigen 45A5; Cancer/testis antigen 45A6; Cancer/testis antigen 45A7; cancer/testis antigen CT45-1; cancer/testis antigen CT45-3; cancer/testis antigen CT45-5; cancer/testis antigen CT45-6; cancer/testis antigen family 45 member A1; Cancer/testis antigen family 45 member A10; cancer/testis antigen family 45 member A3; Cancer/testis antigen family 45 member A4; cancer/testis antigen family 45 member A4/A6; cancer/testis antigen family 45 member A5; Cancer/testis antigen family 45 member A6; Cancer/testis antigen family 45 member A7; cancer/testis antigen family 45 member A-like; cancer/testis antigen family 45, member A1; cancer/testis antigen family 45, member A10; cancer/testis antigen family 45, member A3; cancer/testis antigen family 45, member A4; cancer/testis antigen family 45, member A5; cancer/testis antigen family 45, member A6; CT45; CT45.1; CT45.3; CT45.4; CT45.5; CT45.6; CT45-1; CT45-3; CT45-4; CT455; CT45-5; CT45-6; CT45A1; CT45A10; CT45A3; CT45A4; CT45A5; CT45A6; CT45A7
Gene Symbols CT45A1, CT45A10, CT45A3, CT45A5, CT45A6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CT45A. Recombinant protein control fragment (Product #RP-104372).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102723631, 441519, 441521, 541465, 541466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.