Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CT47B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571075
Description
This target displays homology in the following species: Human: 100%.
CT47B1 is a protein coding gene.
Specifications
CT47B1 | |
Polyclonal | |
Unconjugated | |
CT47B1 | |
Cancer/testis antigen 47.13; cancer/testis antigen 47B; cancer/testis antigen family 147, member B1; cancer/testis antigen family 47, member B1; cancer/testis CT47 family, member 13; CT47.13; CT47A13; CT47B1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
643311 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0C2W7 | |
CT47B1 | |
synthetic peptide directed towards the C-terminal region of human CT47B1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction