Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CT83 Monoclonal Antibody (CL4762)
GREENER_CHOICE

Catalog No. PIMA524711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524711 Supplier Invitrogen™ Supplier No. MA524711
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: QRNTGEMSSNS TALALVRPSS SGLINSNTDN NLAVYDLSRD ILNNFPHSIA RQKRILVNLS MVENKLVELE HTLLSKGFRG ASPHRKST Binds to an epitope located within the peptide sequence STALALVRPSSSGLI as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs - mouse 30%, rat 30%.

CT83 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CT83
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL4762
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CT83
Gene Accession No. Q5H943
Gene Alias Cancer/testis antigen 83; CT83; CXorf61; kita-kyushu lung cancer antigen 1; KKLC1; KK-LC-1
Gene Symbols CT83
Host Species Mouse
Immunogen Recombinant protein fragment of Human CT83 (Product #RP-92344)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203413
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.