Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CT83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA552102 Supplier Invitrogen™ Supplier No. PA552102
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QRNTGEMSSN STALALVRPS SSGLINSNTD NNLAVYDLSR DILNNFPHSI ARQKRILVNL SMVENKLVEL EHTLLSKGFR GASPHRKST Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.

CT83 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CT83
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CT83
Gene Accession No. Q5H943
Gene Alias Cancer/testis antigen 83; CT83; CXorf61; kita-kyushu lung cancer antigen 1; KKLC1; KK-LC-1
Gene Symbols CT83
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CT83. Recombinant protein control fragment (Product #RP-92344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203413
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.