Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CTA Base Control (Cystine Trypticase Agar)

Catalog No. R060570
Click to view available options
Quantity:
20 x 4 mL
100 x 4 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Identify and maintain Neisseria gonorrhoeae and other bacteria using CTA Base Control (Cystine Trypticase Agar).

Identify and maintain Neisseria gonorrhoeae and other bacteria using Thermo Scientific™ Remel™ CTA Base Control (Cystine Trypticase Agar). This agar was first developed by Vera1. CTA Base Control media without carbohydrates can be used for the maintenance of stock cultures while CTA with carbohydrates differentiates fastidious organisms by means of fermentation reactions. Motility can be developed by CTA when the medium is inoculated by a stab line2.

Identify and maintain Neisseria gonorrhoeae and other bacteria using CTA Base Control (Cystine Trypticase Agar).

  • Can be used for maintaining stock cultures
  • Can be used for motility testing
  • Acts as a fermentation control to the CTA medium with carbohydrate to indicate a system failure such as carryover of carbohydrate from a primary isolation medium.
  • Ready to Use: Convenience of prepared media.

This medium can be used for qualitative procedures. CTA Agar consists of cysteine and casein peptone which supply essential nutrients for the growth of fastidious organisms. Neutral-orange color is caused by bacterial deamination of the peptone in the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description CTA Base Control (Cystine Trypticase Agar)
Format 15mm x 103mm Tube
Product Type Agar
Quantity 100 x 4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.