Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTAG1B Antibody (OTI2B6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270485V
Description
CTAG1B Monoclonal specifically detects CTAG1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CTAG1B | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTAG1B | |
Full-length protein expressed in 293T cell transfected with human CTAG1B expression vector. | |
0.1 mL | |
1485.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2B6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction