Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTAG1B Antibody (OTI2B6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147714
Description
CTAG1B Monoclonal specifically detects CTAG1B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CTAG1B | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:5000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1 | |
Mouse | |
18 kDa | |
0.1 mL | |
Primary | |
Human, Monkey | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI2B6 | |
Unconjugated | |
P78358 | |
CTAG1B | |
Full-length protein expressed in 293T cell transfected with human CTAG1B expression vector | |
Immunogen affinity purified | |
RUO | |
1485 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction