Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTAG1B Mouse anti-Human, Clone: UMAB73, liquid, UltraMAB™
SDP

Catalog No. 501684951
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4951 Supplier OriGene Supplier No. UM500061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by CTAG1B is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.

Specifications

Antigen CTAG1B
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UMAB73
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTAG1B
Gene Alias CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1
Gene Symbols CTAG1B
Host Species Mouse
Immunogen Full length human recombit protein of human CTAG1B produced in HEK293T cell
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1485
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.