Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTAG1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577931 Supplier Invitrogen™ Supplier No. PA577931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A375 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by CTAG1B is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTAG1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CTAG1B
Gene Accession No. P78358
Gene Alias Autoimmunogenic cancer/testis antigen NY-ESO-1; cancer antigen 3; cancer/testis antigen 1; cancer/testis antigen 1B; cancer/testis antigen 6.1; CT6.1; CTAG; CTAG1; CTAG1A; CTAG1B; ESO1; l antigen family member 2; LAGE2; LAGE-2; LAGE2A; LAGE2B; New York esophageal squamous cell carcinoma 1; NY-ESO-1
Gene Symbols CTAG1B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CTAG1B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1485
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.