Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTAG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571845
Description
CTAG2 Polyclonal Antibody for Western Blot, IHC (P)
CTAG2 encodes an autoimmunogenic tumor antigen that belongs to the ESO/LAGE family of cancer-testis antigens. This protein is expressed in a wide array of cancers including melanoma, breast cancer, bladder cancer and prostate cancer. This protein is also expressed in normal testis tissue. An alternative open reading frame product of this gene has been described in PMID:10399963. This alternate protein, termed CAMEL, is a tumor antigen that is recognized by melanoma-specific cytotoxic T-lymphocytes. Alternate splicing results in multiple transcript variants.
Specifications
CTAG2 | |
Polyclonal | |
Unconjugated | |
CTAG2 | |
Autoimmunogenic cancer/testis antigen NY-ESO-2; CAMEL; cancer/testis antigen 2; Cancer/testis antigen 6.2; cancer/testis antigen family 6, member 2a; cancer/testis antigen family 6, member 2b; CT2; CT6.2; CT6.2a; CT6.2b; CT62; Ctag2; CTL-recognized antigen on melanoma; ESO2; L antigen family member 1; LAGE1; LAGE-1; LAGE-1a protein; LAGE2B; RGD1561702 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
30848 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O75638 | |
CTAG2 | |
KLH conjugated synthetic peptide between 122-149 amino acids from the Central region of human CTAG2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction