Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTAGE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112107
Description
CTAGE1 Polyclonal Antibody for Western Blot, IP
CTAGE1 (Cutaneous T Cell Lymphoma-Associated Antigen 1) is a Protein Coding gene. Diseases associated with CTAGE1 include Cutaneous T Cell Lymphoma and Lymphoma. An important paralog of this gene is MIA2.
Specifications
| CTAGE1 | |
| Polyclonal | |
| PBS with no preservative | |
| Q96RT6, Q9HC47 | |
| CTAGE-2, CTAGE1 | |
| A synthetic peptide corresponding to the N-terminus of the Human CTAGE1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CTAGE1 | |
| Cancer/testis antigen 21.2; cancer/testis antigen family 21, member 1; cancer/testis antigen family 21, member 2; CT21.1; CT21.2; CTAGE; cTAGE family member 2; CTAGE1; CTAGE-1; CTAGE2; CTAGE-2; cutaneous T-cell lymphoma-associated antigen 1; cutaneous T-cell lymphoma-associated antigen 2; protein cTAGE-2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 64693 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction