Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTAGE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116332
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116332 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116332 Supplier Invitrogen™ Supplier No. PA5116332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTAGE1 Polyclonal Antibody for Western Blot

CTAGE1 (Cutaneous T Cell Lymphoma-Associated Antigen 1) is a Protein Coding gene. Diseases associated with CTAGE1 include Cutaneous T Cell Lymphoma and Lymphoma. An important paralog of this gene is MIA2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTAGE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CTAGE1
Gene Accession No. Q96RT6, Q9HC47
Gene Alias Cancer/testis antigen 21.2; cancer/testis antigen family 21, member 1; cancer/testis antigen family 21, member 2; CT21.1; CT21.2; CTAGE; cTAGE family member 2; CTAGE1; CTAGE-1; CTAGE2; CTAGE-2; cutaneous T-cell lymphoma-associated antigen 1; cutaneous T-cell lymphoma-associated antigen 2; protein cTAGE-2
Gene Symbols CTAGE-2, CTAGE1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 386-414 amino acids from the Central region of human CTAGE1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64693
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.