Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTAGE5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581872 Supplier Invitrogen™ Supplier No. PA581872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEMKLHRKLT VEENYRLEKE EKLSKVDEKI SHATEELETY RKRAKDLEEE LERTIHSYQG QIISHEKKAH DNWLAARNAE RNLNDLRKEN AHNRQKLTET ELKFELLEKD PYALDV Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 95%.

CTAGE5/MIA-2 is a secreted cytokine, and a member of the MIA/OTOR family. Members of this family, which also includes MIA, OTOR, and TANGO, share a Src homology-3 (SH3)-like domain. MIA-2 is predominantly expressed in hepatocytes. Elevated levels of MIA-2 may represent a clinically useful marker for diagnosis of hepatic disease activity and severity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTAGE5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mia2
Gene Accession No. Q96PC5
Gene Alias cTAGE family member 5; CTAGE family member 5 ER export factor; CTAGE family member 5, ER export factor; CTAGE family, member 5; CTAGE5; cutaneous T-cell lymphoma-associated antigen 5; LOW QUALITY PROTEIN: melanoma inhibitory activity protein 2; MEA11; MEA6; melanoma inhibitory activity 2; melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); meningioma-expressed antigen 6/11; MGEA; MGEA11; MGEA6; MIA SH3 domain ER export factor 2; MIA2; protein cTAGE-5
Gene Symbols Mia2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CTAGE5. Recombinant protein control fragment (Product #RP-93697).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4253
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.