Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTAGE5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7516 Supplier Bioss Supplier No. BS6808R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTAGE5 is a tumor-associated antigen.

Specifications

Antigen CTAGE5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mia2
Gene Accession No. Q96PC5
Gene Alias BOS_20386; CTAGE family member 5; CTAGE family member 5 ER export factor; CTAGE family member 5, ER export factor; CTAGE family, member 5; CTAGE5; cutaneous T-cell lymphoma-associated antigen 5; cutaneous T-cell lymphoma-associated antigen 5 homolog; LOC100912115; LOW QUALITY PROTEIN: melanoma inhibitory activity protein 2; MEA11; MEA6; melanoma inhibitory activity 2; Melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); Meningioma-expressed antigen 6/11; MGEA; MGEA11; MGEA6; MIA SH3 domain ER export factor 2; MIA2; protein cTAGE-5
Gene Symbols Mia2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CTAGE5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4253
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.