Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTAGE6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549461

Catalog No. PA549461


Only null left
Add to Cart

Description

Description

CTAGE6 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTAGE6
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q86UF2
CTAGE6
KLH conjugated synthetic peptide between 581-615 amino acids from the C-terminal region of human CTAGE6P.
80 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
CTAGE6
cTAGE family member 6; CTAGE family, member 6; CTAGE family, member 6, pseudogene; CTAGE6; CTAGE6P; cutaneous T-cell lymphoma-associated antigen 6; protein cTAGE-6; putative protein cTAGE-6
Rabbit
Antigen affinity chromatography
RUO
340307
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.