Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CtBP1 Antibody (4D6), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174040AF594
Description
CtBP1 Monoclonal specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CtBP1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CTBP1 | |
Recombinant human CTBP1 (1-440aa) purified from E. coli | |
Primary | |
Human | |
IgG2b κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
4D6 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 | |
Mouse | |
100 μL | |
1487 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction