Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CtBP1 Antibody (4D6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174040H
Description
CtBP1 Monoclonal specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| CtBP1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CTBP1 | |
| Recombinant human CTBP1 (1-440aa) purified from E. coli | |
| 100 μL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 1487 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4D6 | |
| Western Blot, ELISA | |
| BARS, brefeldin A-ribosylated substrate, CTBP, CtBP1, C-terminal binding protein 1, C-terminal-binding protein 1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.158, MGC104684 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human CTBP1 mAb, clone 4D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CTBP1 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction