Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CtBP1/CtBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5106258

Description
Antibody detects endogenous levels of total CtBP1/2.
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications
CtBP1/CtBP2 | |
Polyclonal | |
Unconjugated | |
CTBP2 | |
50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; AA407280; Bars; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; Ctbp1; CtBP1-L; CtBP1-S; CTBP2; Ctbp3; CtBP3/BARS; C-terminal binding protein 1; C-terminal binding protein 2; C-terminal-binding protein 1; C-terminal-binding protein 2; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; ribeye | |
Rabbit | |
Affinity chromatography | |
RUO | |
13016, 13017, 1487, 1488, 29382, 81717 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O88712, P56545, P56546, Q13363, Q9EQH5, Q9Z2F5 | |
CTBP1, CTBP2 | |
A synthesized peptide derived from human CTBP1(Accession Q13363), corresponding to amino acid residues T132-L182. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction