Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CTBP1-DT Antibody (OTI2C6), FITC, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272333F
Description
CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| CTBP1-DT | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CTBP1-DT | |
| Recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 (NP_443093) produced in E.coli. | |
| 0.1 mL | |
| 92070.0 | |
| Store at 4°C in the dark. | 
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2C6 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CTBP1 antisense RNA 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            