Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTBP1-DT Antibody (OTI4F2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245585
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry
CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
C4orf42 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100 | |
CTBP1 antisense RNA 1 | |
Mouse | |
19.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
OTI4F2 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CTBP1-DT | |
Human recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42(NP_443093) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
92070 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction