Learn More
Invitrogen™ CtBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595014
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Testis Tissue, Rat Ovary Tissue, U87 Cell, SW620 Cell, HT1080 Cell, COLO32 Cell. IHC: Rat Brain Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications
CtBP1 | |
Polyclonal | |
Unconjugated | |
CTBP1 | |
50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13016, 1487, 29382 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O88712, Q13363, Q9Z2F5 | |
CTBP1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human CtBP1 (425-440aa QTVKPEADRDHASDQL). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.