Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CtBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595252
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595252 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595252 Supplier Invitrogen™ Supplier No. PA595252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human SH-SY5Y whole cell, human MOLT-4 whole cell, human MCF-7 whole cell, rat PC-12 whole cell, mouse SP2/0 whole cell. IHC: Mouse Thymus tissue, Rat Spleen tissue, Human Tonsil tissue. ICC/IF: U20S cell, T47D cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CtBP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene CTBP1
Gene Accession No. O88712, Q13363, Q9Z2F5
Gene Alias 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684
Gene Symbols CTBP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human CtBP1 (409-440aa HGLPPVAHPPHAPSPGQTVKPEADRDHASDQL).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13016, 1487, 29382
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.