Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CtBP1 Polyclonal Antibody
CtBP1 Polyclonal Antibody
Supplier: Thermo Scientific PA595874
Description
CtBP1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
CtBP1 | |
Polyclonal | |
Unconjugated | |
CTBP1 | |
50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13016, 1487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.78 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88712, Q13363 | |
CTBP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-250 of human CtBP1 (NP_0013191). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction