Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTBP2 Antibody (PCRP-CTBP2-1A9), Janelia Fluor™ 549 - C-terminus, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308549JF549
Description
CTBP2 Monoclonal antibody specifically detects CTBP2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
CTBP2 | |
Monoclonal | |
Janelia Fluor 549 | |
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2 | |
Recombinant full-length human CTBP2 protein (Uniprot: P56545) | |
100 μg | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
1488 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-CTBP2-1A9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction