Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTBP2 Antibody (SR1810), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32241550UL
Description
CTBP2 Monoclonal antibody specifically detects CTBP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
CTBP2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1810 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2 | |
A synthesized peptide derived from CTBP2 (Uniprot #: P56545) | |
50 μg | |
Cell Cycle and Replication, Wnt Signaling Pathway | |
1488 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction