Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTBP2 Mouse anti-Human, Clone: PCRP-CTBP2-1A9, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen CTBP2
Clone PCRP-CTBP2-1A9
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123052
SDP
View Documents
Novus Biologicals
NBP30778920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

CTBP2 Monoclonal specifically detects CTBP2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

CTBP2
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Cell Cycle and Replication, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
1488
IgG2b
Protein A or G purified
PCRP-CTBP2-1A9
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
CtBP2, C-terminal binding protein 2, 10ribeye, C-terminal-binding protein 2
Recombinant full-length human CTBP2 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.