Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CtBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595843 Supplier Invitrogen™ Supplier No. PA595843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, COS-7, BxPC-3, NIH/3T3, Mouse brain, Rat brain Immunogen sequence: SEPFSFAQGP LKDAPNLICT PHTAWYSEQA SLEMREAAAT EIRRAITGRI PESLRNCVNK EFFVTSAPWS VIDQQAIHPE LNGATYRYPP GIVGVAPGGL PAAMEGIIPG GIPVTHNLPT VAHPSQAPSP NQPTKHGDNR EHPNEQ.

This gene produces alternative transcripts encoding two distinct proteins. One protein is a transcriptional repressor, while the other isoform is a major component of specialized synapses known as synaptic ribbons. Both proteins contain a NAD+ binding domain similar to NAD+-dependent 2-hydroxyacid dehydrogenases. A portion of the 3' untranslated region was used to map this gene to chromosome 21q21. 3; however, it was noted that similar loci elsewhere in the genome are likely. Blast analysis shows that this gene is present on chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CtBP2
Applications ELISA, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CTBP2
Gene Accession No. P56545, P56546, Q9EQH5
Gene Alias AA407280; CTBP2; C-terminal binding protein 2; C-terminal-binding protein 2; D7Ertd45e; gene trap ROSA b-geo 6; Gtrgeo6; Ribeye; RP11-59C5.2
Gene Symbols CTBP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human CTBP2 (NP_0013201).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13017, 1488, 81717
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.