Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTBS Polyclonal Antibody

Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | CTBS |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119085
![]() |
Invitrogen™
PA5119085 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122060
|
Invitrogen™
PA5122060 |
20 μL |
Each for $209.50
|
|
|||||
Description
CTBS Polyclonal Antibody for Western Blot, IHC (P)
Involved in the degradation of asparagine-linked glycoproteins. Hydrolyze of N-acetyl-beta-D-glucosamine (1-4)N-acetylglucosamine chitobiose core from the reducing end of the bond, it requires prior cleavage by glycosylasparaginase.Specifications
CTBS | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
CTBS | |
2210401K11Rik; chitobiase; chitobiase, di-N-acetyl-; CTB; CTBdi-N-acetylchitobiase; CTBS; Di-N-acetylchitobiase | |
CTBS | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q01459 | |
1486 | |
Recombinant protein Di-N-Acetyl Chitobiase. The antigen corresponds to amino acid range 47-385 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title