Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTC-534A2.2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24956425UL
Description
CTC-534A2.2 Polyclonal antibody specifically detects CTC-534A2.2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CTC-534A2.2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
REV7-interacting novel NHEJ regulator 1, RINN1; CTC-534A2.2, shield complex subunit 3, shieldin complex subunit 3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: HDAKSHSYDCTVDLLEFQPSLKKQHLTWSHTLKEQTNSGNLGKQSEKGKQHKRRSWSISLPSNNCT | |
25 μL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction