Learn More
CTCF Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP259228
Catalog No. NBP259228
Only null left
Add to Cart
Description
Description
CTCF Polyclonal specifically detects CTCF in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Chromatin Immunoprecipitation Sequencing, Knockdown Validated.
Specifications
Specifications
CTCF
Polyclonal
Western Blot 1:1000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200, Chromatin Immunoprecipitation (ChIP) 1 ug/IP, Chromatin Immunoprecipitation Sequencing, Knockdown Validated
11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
CTCF
The exact immunogen is propietary information.
50 μg
Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
10664
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only