Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTCF Monoclonal Antibody (KT71)
GREENER_CHOICE

Catalog No. PIMA549543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549543 Supplier Invitrogen™ Supplier No. MA549543
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This product was used in an Indirect ELISA.

Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTCF
Applications Western Blot
Classification Monoclonal
Clone KT71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CTCF
Gene Accession No. P49711, Q61164
Gene Alias 11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283)
Gene Symbols CTCF
Host Species Rat
Immunogen Recombinant Mouse Ctcf.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10664, 13018
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.