Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTCF Antibody (BLR041F), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP276420
Description
CTCF Monoclonal specifically detects CTCF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
CTCF | |
Monoclonal | |
Unconjugated | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide | |
CTCF | |
Immunogen sequence between amino acids 650 and 700 | |
100 μL | |
10664 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
BLR041F | |
Western Blot 1:1000, Simple Western 1:10 - 1:250, Flow Cytometry Fixed in 4% formaldehyde and permeabilized with 90% methanol. 1 μl, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 20 ul/1mg lysate, Immunohistochemistry-Paraffin 1:100- 1:500, Immunocytochemistry 1:100 - 1:500 | |
11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF | |
Rabbit | |
Antigen Affinity-purified | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction