Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTCFL Monoclonal Antibody (KT72)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549545
Description
This product was used in an Indirect ELISA.
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
Specifications
CTCFL | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
A2APF3 | |
Ctcfl | |
Recombinant Mouse Ctcfl. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Western Blot | |
KT72 | |
Unconjugated | |
Ctcfl | |
BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CCCTC-binding factor like; CT27; CTCF paralog; CTCF T; CTCFL; CTCF-like protein; CTCF-T; dJ579F20.2; HMG 1L1; HMG-1L1; HMGB1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; OTTMUSG00000016680; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; transcriptional repressor CTCFL; Zinc finger protein CTCF-T | |
Rat | |
Protein G | |
RUO | |
664799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction