Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CTCFL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551584
Description
Immunogen sequence: DGVCREKDHR SPSELEAQRT SGAFQDSVLE EEVELVLAPS EESEKYILTL QTVHFTSEAV ELQDMSLLSI QQQEGVQVVV QQPGPGLLWL EEGPRQSLQQ CVAISIQQEL YSPQEMEVLQ FHALEENVMV ASEDSKLAVS Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 36%.
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
Specifications
| CTCFL | |
| Polyclonal | |
| Unconjugated | |
| Ctcfl | |
| BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CCCTC-binding factor like; CT27; CTCF paralog; CTCF T; CTCFL; CTCF-like protein; CTCF-T; dJ579F20.2; HMG 1L1; HMG-1L1; HMGB1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; OTTMUSG00000016680; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; transcriptional repressor CTCFL; Zinc finger protein CTCF-T | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NI51 | |
| Ctcfl | |
| Recombinant protein corresponding to Human CTCFL. Recombinant protein control fragment (Product #RP-96690). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction