Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDP1/FCP1 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen CTDP1/FCP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-155-9444
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301172AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
Specifications

Specifications

CTDP1/FCP1
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
CCFDN, FCP1, CTD of POLR2A, phosphatase of, subunit 1, RNA polymerase II subunit A C-terminal domain phosphatase, TFIIF-associating CTD phosphatase 1, serine phosphatase FCP1a, transcription factor IIF-associating CTD phosphatase 1
Between 911 and 961
Primary
4° C
CTDP1
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q7TSG2, Q9Y5B0
67655, 9150
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.